VWR® Antimagnetic Forceps
Supplier: VWR International
82027-400
82027-400CS 2913.05 USD
82027-400
VWR® Antimagnetic Forceps
Forceps
Antimagnetic forceps are constructed from anti-magnetic stainless steel.
  • Good resistance to corrosion
  • Autoclavable
  • For general-purpose use
Forceps feature an overall length of 114 mm (4.5") and are available in a variety of unit sizes, from individual forceps to bulk packages.