VWR® Tongs, PTFE-Coated
Supplier: VWR International
82027-376
82027-376CS 1550.58 USD
82027-376
VWR® Tongs, PTFE-Coated
Tongs
Designed to handle hot, cold, corrosive, or sticky materials without scratching or damaging container exteriors.
  • Stainless steel construction
  • PTFE coating
  • Nonsticking and non-corrosive
Contoured jaw enables the handling of crucibles and other objects. Tongs are used for handling chemically active materials.