VWR® Vacuum Bulb
Supplier: VWR International
82024-578
82024-578CS 3691.74 USD
82024-578
VWR® Vacuum Bulb
Samplers Gas Samplers
Designed for use in gas sampling or for pumping gas through apparatus.
  • Heavy-walled design
  • Ideal for use in gas sampling
  • Supplied independent of tubing
Heavy-walled bulbs feature two rubber valves with PVC connections. Bulb capacity is 85 mL. Not supplied with tubing.