VWR® Large Pipet Drawer Organizer
Supplier: VWR International
82024-548
82024-548CS 907.41 USD
82024-548
VWR® Large Pipet Drawer Organizer
Trays/Dishes/Pots Drawer Organization Trays
This rugged polystyrene drawer organizer is ideal for storing a variety of glass or plastic pipets and supplies
  • Rugged polystyrene construction
  • Convenient compact size
  • Multiple compartments of varying size
The convenient size fits into standard laboratory drawers or on benches, tabletops, or shelves. Each tray features three compartments of varying size.
Items are kept organized and easily accessible.
First compartment: 43 x 11 cm (17 x 45/16")
Second and third compartment: 43 x 8 cm (17 x 31/8")