VWR® 8-Well Centrifuge Tube Rack
Supplier: VWR International
82024-452
82024-452CS 6446.32 USD
82024-452
VWR® 8-Well Centrifuge Tube Rack
Racks Tube Racks
Rack is ideal for holding up to eight 50 mL conical centrifuge tubes and features a slotted bottom for secure handling.
  • Autoclavable
  • Resistant to chemicals
Rack can also accommodate other types and sizes of tubes. Tubes sit in 30mm square holes. Epoxy-coated steel wire rack is chemical-resistant and autoclavable.