Glass Tubing, United Scientific Supplies
Supplier: United Scientific Supplies
GT-L-06 GT-L-07 GT-L-09 GT-L-25 GT-M-30 GT-H-75
82024-092CS 420.3 USD
82024-092 82024-094 82024-096 82024-098 82024-100 82024-102
Glass Tubing, United Scientific Supplies
Tubing Transfer Tubing
Borosilicate glass tubing.
  • Various diameters
Choice of thin, standard, or heavy wall tubing.