Griffin Beakers, Low Form, United Scientific Supplies
Supplier: United Scientific Supplies
BG1000-3000
82024-074CS 1032.85 USD
82024-074
Griffin Beakers, Low Form, United Scientific Supplies
Beakers
Borosilicate glass beakers are ideal for heating because of an optimum balance between thermal resistance and mechanical resistance achieved from controlled wall thickness
  • Size ranges from 5 ml to 20000 ml
  • Tolerance for beakers is approximately +/- 5%
  • Low-expansion borosilicate glass
All beakers feature pour-off spouts. The graduated beakers feature white graduation marks.