Iodoacetamide

Supplier: G-Biosciences
Warning

RC-150 786-228
71003-536EA 452.68 USD
71003-536 82023-298
Iodoacetamide
Iodoacetamide

G-Biosciences offers proteomic grade iodoacetamide in two bulk quantities (5 or 25 grams) or in OneQuant™ format. G-Biosciences' OneQuant™ Iodoacetamide are single aliquots of iodoacetamide that eliminate the need for weighing, preventing loss of reagent

Formula: ICH₂CONH₂
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 91…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR