Iodoacetamide, OneQuant™

Supplier: G-Biosciences
Warning

786-078
82023-296EA 240.37 USD
82023-296
Iodoacetamide, OneQuant™
Iodoacetamide

OneQuant™ iodoacetamide are single aliquots of iodoacetamide that eliminate the need for weighing, preventing loss of reagent and saving time. Add 150μl water to a single tube to generate a 0.5M iodoacetamide solution. Supplied with 24 individual tubes.

Formula: ICH₂CONH₂
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 91…94 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008028
CAS Number: 144-48-9
EINECS: 205-630-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR