Ethidium bromide 10 mg/ml in aqueous solution

Supplier: G-Biosciences
Warning

R034
82023-294EA 132.63 USD
82023-294
Ethidium bromide 10 mg/ml in aqueous solution
Ethidium bromide
Formula: C₂₁H₂₀BrN₃
MDL Number: MFCD00011724
CAS Number: 1239-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR