FOCUS™ Mitochondria, G-Biosciences
Supplier: G-Biosciences
786-022
82023-258EA 477.04 USD
82023-258
FOCUS™ Mitochondria, G-Biosciences
Protein Extraction Protein Extraction Kits
Specifically designed for the isolation of intact mitochondria from cultured mammalian cells
This kit is highly adaptable for use with animal tissues and other sources of mitochondria. The kit is designed for 50 preps, where one prep is equivalent to 2x10^7 mammalian cells or 100 mg mammalian tissue.
This kit allows for the fast and efficient fractionation of the cytoplasm of cultured mammalian cells into an enriched fraction of mitochondria. The majority (>90%) of the mitochondria have intact inner and outer membranes and therefore retain their functionality.
Ordering information: Accessories available.