OmniTemplate™ for Single Tube Preparation of PCR Template DNA, G-Biosciences
Supplier: G-Biosciences
786-013
82023-256EA 306.37 USD
82023-256
OmniTemplate™ for Single Tube Preparation of PCR Template DNA, G-Biosciences
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
G-Biosciences' OmniTemplate™ is specifically designed for the rapid isolation of a DNA template for polymerase chain reaction (PCR) analysis from mammalian tissue samples, blood and cell cultures
G-Biosciences' OmniTemplate™ method involves isolation of nuclei and their subsequent digestion with LongLife™ Proteinase K in the same tube the samples were collected in. The isolated nuclei are digested to release template DNA in a buffer compatible with PCR and other downstream applications. The OmniTemplate™ samples can be directly used in PCR and other applications without further manipulations. One prep is approximately 1-10 mg tissue.
This single tube method produces a high concentration, ready-to-use supply of DNA template for microanalysis, genotyping and a multitude of other applications.
Ordering information: Accessories available.