Calcium chloride 1 M in aqueous solution

Supplier: G-Biosciences
Warning

Synonyms: Chloro calcium

R040
82023-114EA 56.63 USD
82023-114
Calcium chloride 1 M in aqueous solution
Calcium chloride solutions
Formula: CaCl₂·2H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR