D-(-)-Sorbitol solution 1 M

Supplier: G-Biosciences

R038
82023-110EA 51.45 USD
82023-110
D-(-)-Sorbitol solution 1 M
D(-)-Sorbitol

Quality DNase and RNase free reagent solution for yeast research.

Formula: HOH₂C(CH(OH))₄CH₂OH
Storage Temperature: Refrigerator
MDL Number: MFCD00004708
CAS Number: 50-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR