Ammonium acetate solution 5M

Supplier: G-Biosciences

R012
82023-100EA 58.95 USD
82023-100
Ammonium acetate solution 5M
Ammonium acetate
Formula: H₃CCOONH₄
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR