Magnesium chloride solution 1 M

Supplier: G-Biosciences

R004
82023-086EA 56.63 USD
82023-086
Magnesium chloride solution 1 M
Magnesium chloride solutions
Formula: MgCl₂·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR