HOOK™ Photoreactive Biotin Reagent for Highly Efficient Protein Labeling, G-Biosciences
Supplier: G-Biosciences
BG-20
82022-912EA 498.78 USD
82022-912
HOOK™ Photoreactive Biotin Reagent for Highly Efficient Protein Labeling, G-Biosciences
Protein Linking and Labeling Systems
The use of biotin for non-radioactive labeling of proteins and nucleic acids has now become an increasingly popular technique in life science research
Photoreactive agents, on exposure to ultraviolet light, become active and bind non-specifically with neighboring molecules. These reagents are suitable for labeling molecules that do not contain easily reactable functional groups.
G-Biosciences' HOOK™-Psoralen-PEO-Biotin, a photoreactive reagent, reacts and labels nucleic acids and protein molecules. When reacted with nucleic acids, it cross-links with pyrimidine bases. Cross-linking does not interfere with hybridization applications.
Several factors must be considered when coupling a biotin reagent to a protein to ensure a successful reaction. The primary consideration is the selection of the biotinylation reagent itself. G-Biosciences offers a wide range of biotin reagents with variations in their reactive groups, spacer arm lengths, solubility, membrane permeability and reversibility. All of these factors must be considered and are dependent on your protein/peptide.
Ordering information: Accessories included.