SATA (N-Succinimidyl S-Acetylthioglycolate)

Supplier: G-Biosciences

Synonyms: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

BC96
82022-864EA 275.11 USD
82022-864
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)

G-Biosciences' SATA reacts with primary amines, adding protected sulfhydryl residues.


G-Biosciences' SATA is soluble in DMSO and the sulfhydryl group can be used in coupling reactions.


G-Biosciences' SATA is offered as a 100 milligram supply. Formulation: N-Succinimidyl S-acetylthioacetate. Molecular Formula: C₈H₉NO₅S.


Ordering information: Accessories available.

Formula: C₈H₉NO₅S
MW: 231.22 g/mol
MDL Number: MFCD00036891
CAS Number: 76931-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR