SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Supplier: G-Biosciences

BC20
82022-808EA 337.86 USD
82022-808
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)

Succinimidyl 4-(N-maleimidomethyl) cyclohexane-1-carboxylate Heterobifunctional Cross Linkers.


  • Ideal for enzyme-antibody crosslinking

Formula: C₁₆H₁₈N₂O₆
MDL Number: MFCD00009634
CAS Number: 64987-85-5

Order Now

Specification Test Results

Spacer Arm (Å) 11.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable Yes
Water Soluble No
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR