PMPI (4-Maleimidophenyl isocyanate)

Supplier: G-Biosciences
Danger

BC18
82022-804EA 410.11 USD
82022-804
PMPI (4-Maleimidophenyl isocyanate)
PMPI (4-Maleimidophenyl isocyanate)

N-(ρ-Maleimidophenyl) isocyanate Heterobifunctional Cross Linkers.

Formula: C₁₁H₆N₂O₃
MDL Number: MFCD00270900
CAS Number: 123457-83-0

Order Now

Specification Test Results

Spacer Arm (Å) 8.7
Reactive Toward Sulfhydryl + Hydroxyl
Membrane Permeable Not tested
Water Soluble No
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR