Botanical Garden Blot, G-Biosciences
Supplier: G-Biosciences
BB01
82022-768EA 1112.5 USD
82022-768
Botanical Garden Blot, G-Biosciences
Blotting Kits
G-Biosciences' Botanical Garden Blot is prepared using total protein extracted from a variety of 5-10 day old, whole plant tissues.
G-Biosciences' premade blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel, along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
G-Biosciences' premade blots are ready to be blocked and probed with the antibodies of choice. Note: We recommend that customers contact our technical department for the latest information on the blots as G-Biosciences reserves the right to change the blot profile due to the availability of GenLysate™.
Ordering information: Accessories available.