FOCUS™ PhosphoRich™, G-Biosciences
Supplier: G-Biosciences
786-255
82022-696EA 862.6 USD
82022-696
FOCUS™ PhosphoRich™, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' FOCUS™ PhosphoRich™ is a ready-to-use kit that enriches phosphorylated proteins and phosphopeptides from complex biological samples
The kit contains spin columns that have a phosphoprotein binding resin with a binding capacity of ~20mg phosphorylated ovalbumin per column. The resin columns supplied with the kit can be reused, if regenerated and stored properly.
Ordering information: Accessories available.