FOCUS™ Membrane Proteins, G-Biosciences
Supplier: G-Biosciences
786-249
82022-676EA 688.09 USD
82022-676
FOCUS™ Membrane Proteins, G-Biosciences
Protein Extraction Protein Extraction Kits
G-Biosciences' FOCUS™ Membrane Proteins is a rapid and highly reproducible method for preparation of membrane or hydrophobic proteins from biological samples for 2D-gel analysis or other applications
The kit is provided with reagents necessary for extraction of membrane proteins and their subsequent preparation for isoelectric focusing or 2D gel resolution for improved spot resolution.
G-Biosciences' FOCUS™ Membrane Proteins kit is designed for >50 preps, where 1 prep is one of the following: 100 mg mammalian tissue, 50μL wet animal cell pellet, 50μL wet yeast pellet, 50μL wet bacteria pellet, or 250 mg plant tissue.
Membrane proteins are extracted with a single step phase partition, with an efficiency greater than 90% with minimal cross-contamination from hydrophilic proteins.