Nuclear and Cytoplasmic Extraction Kit, G-Biosciences
Supplier: G-Biosciences
786-182
82022-600EA 414.58 USD
82022-600
Nuclear and Cytoplasmic Extraction Kit, G-Biosciences
Buffers Extraction Buffers
For the enrichment of cytoplasmic and nuclear fractions from cultured cells and tissues
This kit is based on organic buffers and contains a proprietary combination of various salts and agents. The kit is provided with reagents for solubilization of the nuclear fraction. This kit is designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100mg mammalian tissue. This kit is designed for >50 preps, depending on the tissue used, where one prep is 20x10^6 mammalian cells or 100 mg mammalian tissue.
The kit generates a cleaner separation of cytoplasmic proteins from nuclear proteins and is ideal for expression and transport studies.