Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), OneQuant™

Supplier: G-Biosciences
Warning

Synonyms: Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulphonate, Sulphosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sodium 1-((4-((2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl)cyclohexane-1-carbonyl)oxy)-2,5-dioxopyrrolidine-3-sulfonate, Sulpho-SMCC Sodium Salt, SMCC, Water-Soluble, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulpho-N-hydroxysuccinimide ester sodium salt, sulfo-SMCC Sodium Salt, sulpho-SMCC, 4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 3-sulfo-N-hydroxysuccinimide ester sodium salt, Sulfosuccinimidyl-trans-4-(N-maleimidomethyl)cyclohexane-1-carboxylate, Sulfo-SMCC

786-082
82022-426PK 233.56 USD
82022-426
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt), OneQuant™
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt)

Ideal for enzyme-antibody crosslinking.


  • No weighing required


OneQuant™ Sulfo SMCC: Single use vials to minimize waste.

Formula: C₁₆H₁₈N₂O₉S
MW: 436.37 g/mol
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now

Specification Test Results

Spacer Arm (Å) 11.6
Reactive Toward Primary Amine + Sulfhydryl
Membrane Permeable No
Water Soluble Yes
Cleavable/ Reversible No


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR