Dexamethasone 100 mM

Supplier: G-Biosciences

API-04
82021-602EA 189.57 USD
82021-602
Dexamethasone 100 mM
Dexamethasone

Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).


G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.


Dexamethasone is a potent and stable glucocorticoid steroid hormone that is often used as an anti-inflammatory and anti-rheumatic drug.
The mechanism of action is not fully understood, however it is thought that it may inhibit prostaglandin synthesis.

Formula: C₂₂H₂₉FO₅
MDL Number: MFCD00064136
CAS Number: 50-02-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR