Actinomycin D 10 mmol/l in aqueous solution

Supplier: G-Biosciences

API-01
82021-596EA 189.8 USD
82021-596
Actinomycin D 10 mmol/l in aqueous solution
Actinomycin D

Apoptosis is commonly defined as programmed mechanism of cell death and is a highly regulated process that can be initiated by a variety of internal and external stimuli (inducers).


  • Supplied as 50μl 10mM solution


G-Biosciences’ Apoptosis Inducers are ready-to-use, high quality solutions for inducing apoptosis in various cell types.


Actinomycin D inhibits transcription by binding to DNA at the transcription initiation complex and therefore preventing elongation by the RNA polymerase.

Formula: C₆₂H₈₆N₁₂O₁₆
MDL Number: MFCD00005033
CAS Number: 50-76-0
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR