N,N-Dimethylformamide, anhydrous

Supplier: G-Biosciences
Danger

BKC-16
82021-450EA 114.38 USD
82021-450
N,N-Dimethylformamide, anhydrous
N,N-Dimethylformamide

Anhydrous DMF [N,N-Dimethylformamide (HCON(CH3)2)]. Suitable for cross-linking, protein modification and biotinylation reaction applications.

Formula: HCON(CH₃)₂
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.949 g/cm³ (20 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ac5c5e9758b6c40ccb0cf127b6a6e1c6d608c64&vid=9a5d4e579e195193602c20d3cb39c89ac9c92cbf&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR