Silver D-Stain™ Nucleic Acid & Protein Destain, G-Biosciences
Supplier: G-Biosciences
786-199
82021-352EA 304.37 USD
82021-352
Silver D-Stain™ Nucleic Acid & Protein Destain, G-Biosciences
Electrophoresis Reagents Stain Removal Reagents
G-Biosciences' Silver D-Stain™ can be used for reducing the staining of silver stained gels and revealing over-stained bands
G-Biosciences' Silver D-Stain™ allows you to destain gels completely and restain, or follow other downstream applications such as Western transfer or sequencing.
G-Biosciences' Silver D-Stain™ is suitable for destaining and restaining both silver stained protein gels and nucleic acid gels.
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.