EKONO™ Hybridization Buffer, G-Biosciences
Supplier: G-Biosciences
786-160
82021-338EA 205.57 USD
82021-338
EKONO™ Hybridization Buffer, G-Biosciences
Buffers Electrophoresis Buffers Hybridization Buffers
A high performance hybridization buffer system providing researchers with unprecedented simplicity, reliability, accuracy, and cost saving for hybridization
Protocol involves incubation of blots with denatured probes in EKONO™ hybridization buffer followed by washing of the blot to remove excess unhybridized probes. Easy-to-use EKONO™ provides high resolution and sensitivity.
Suitable for Southern/Northern or Southwestern hybridization analysis, hybridization with oligonucleotides or other types of probes, and for hybridization screening of gene libraries.
EKONO™ hybridization buffer is suitable for all types of probes