VWR® Safety Brix Sugar Scale Combined Form Thermo-Hydrometers
Supplier: VWR International
82018-070 82021-182 82021-184 82021-188 82021-190
82018-070EA 119.56 USD
82018-070 82021-182 82021-184 82021-188 82021-190
VWR® Safety Brix Sugar Scale Combined Form Thermo-Hydrometers
Hydrometers
This wide range of combined-form hydrometers are used in the food and beverage, and other industries to measure % sucrose by weight in various ranges from 0° to 61° Brix, and include a convenient, built-in thermometer with a temperature scale of 0/50°C.
  • Conform to the accuracy requirements for ASTM thermo-hydrometers
  • 381 mm (15") long
  • Lead-free ballast and glass
  • Temperature standardization: 20°C
  • Thermometer contains mercury-free, blue spirit organic liquid fill
  • Individually serialized
  • Hand-inspected by U.S. quality team
  • Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST
Each hydrometer conforms to the accuracy requirements for ASTM thermo-hydrometers. Hydrometers include multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.
Caution: Combined form hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures. Enclosed thermometers should be used as a general reference only. For temperature critical applications, a calibrated standard thermometer is recommended.