VWR® Laboratory Hygrometer/Thermometer
Supplier: VWR International
61505-0011
82021-180EA 106.39 USD
82021-180
VWR® Laboratory Hygrometer/Thermometer
Hygrometers Analog Hygrometers
Durable hygrometer/thermometer is encased in stainless steel and can withstand conditions in incubators, refrigerators, and freezers.
  • Stainless steel enclosure
  • Durable for withstanding incubators, freezers, and refrigerators
  • Easy-to-read dials
A black temperature dial and a red humidity dial are set against a white background. Complete with hole for wall mounting.