VWR® Dual Zone Electronic Thermometer
Supplier: VWR International
61506-0211
82021-166EA 172.09 USD
82021-166
VWR® Dual Zone Electronic Thermometer
Hygrometers Digital Thermo-Hygrometers
Feature a waterproof temperature probe, maximum/minimum temperature memory, temperature alert setting, 0.1 resolution, one AAA battery, flip out stand.
  • Waterproof temperature probe
  • Multi-language instructions
Supplied with a multi-language (English, French, German, Italian, Portuguese and Spanish) instructions and Statement of Traceability indicating accuracy traceable to NIST and DKD/PTB.
All thermometers are tested and calibrated in an exclusive triple accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST and DKD/PTB.
Certifications: CE marked. RoHS compliant.