Grafco® Tincture Of Green Soap U.S.P.
Supplier: GF HEALTH PRODUCTS INC. TE
3375
82020-142CS 192.89 USD
82020-142
Grafco® Tincture Of Green Soap U.S.P.
Skincare Soaps and Skin Cleansers
Ideal for removal of dried blood and protein soils from skin, scalp, and hands.
  • Distinctive oil of lavender scent
  • Made in the USA
This vegetable-based alkaline detergent is made from pure vegetable oil, glycerin, and lavender oil. It is 30% alcohol by volume.