Kimtech Prep™ Scottpure™ Critical Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
06151
82019-168CS 671.3 USD
82019-168
Kimtech Prep™ Scottpure™ Critical Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
Kimtech Prep™ Scottpure™ Wipers are designed for wiping tasks where precision and reliability are essential.
  • Engineered for excellent absorbency and low-lint wiping
  • Wipers are solvent resistant, with no binders or surfactants
Rayon/polyester spunlace material for cloth-like softness. Applications include aerospace, clean manufacturing areas, DVD manufacturing, and printing.