VWR® Standard Standard Sealed and Septum Caps
Supplier: VWR International
SCPC38
82013-158CS 301.61 USD
82013-158 82013-160
VWR® Standard Standard Sealed and Septum Caps
Closures Bottle Closures
Caps are manufactured from medical grade, FDA approved, high-density polypropylene.
  • High-density polypropylene construction
  • Reusable and recyclable
  • Autoclavable
  • Supplied nonsterile
They are autoclavable, reusable, and recyclable
Septum caps are available with a gray butyl rubber insert. They are ideal for injectables or containment of pathogens.
VWR® polycarbonate media bottles, vials, and culture tubes are supplied with a standard seal cap. Blue is the standard color for all caps, although custom colors are available upon request.
All caps are linerless with an integrally-molded seal ring to ensure leakproof performance. They are manufactured in a Class 10K cleanroom environment and are supplied nonsterile.