VWR® Pipet Tip Disposal Box
Supplier: VWR International
HS234634V
82010-754EA 203.48 USD
82010-754
VWR® Pipet Tip Disposal Box
Boxes Waste Disposal Boxes
Box features a flip-top port cover for convenient one-handed disposal of used pipet tips.
  • Thick acrylic construction
  • Protects against beta radiation
  • Large, hinged lid
  • Easy disposal of used pipet tips
Eliminates any need to handle pipet tips used with hazardous fluids. The 10mm (3/8") thick acrylic provides protection from beta radiation. Large, hinged lid allows easy removal of contents.