VWR® Cube Racks
Supplier: VWR International
82010-746
82010-746CS 2237.75 USD
82010-746
VWR® Cube Racks
Racks Tube Racks
These polypropylene racks can slide together in multiple orientations to work with different-sized tubes at the same time.
  • Polypropylene fabrication
  • Autoclavable
Each cube rack has four sides that hold either four 50 mL tubes, ten 15 mL conical tubes, twelve 12x75mm or 12x100mm tubes, or sixteen 1.5–2.0 mL microcentrifuge tubes.
Ordering information: Assorted package includes one each of blue, green, pink, yellow, and orange.