Monoject™ Piston Syringes, Covidien
Supplier: Cardinal Health
8881114055
82007-802CS 375.04 USD
82007-802 53498-474
Monoject™ Piston Syringes, Covidien
Syringes Syringes, Medical
Nonsterile, latex-free syringes feature bold dual graduations: 140mL in 5mL increments and 5oz
in 0.25oz. increments. Autoclavable.