KLEENGUARD® A40 Liquid and Particle Protection Sleeve Protectors, Kimberly-Clark Professional®
Supplier: Kimberly-Clark
44480
82007-672CS 194.86 USD
82007-672
KLEENGUARD® A40 Liquid and Particle Protection Sleeve Protectors, Kimberly-Clark Professional®
Disposable Garments
Sleeve protectors are manufactured from breathable, microporous film laminate, which is an excellent particle barrier (100% dry particle holdout between 0.3–0.5µm)
Fabric also passes ASTM F1670 testing for penetration of blood and bodily fluids, for added protection. Sleeves offer a better liquid barrier, better particulate barrier, and are lower in lint than leading competitors. NFPA 99 Compliant Antistatic Material. Sleeves are 45.7cm (18") long and feature elastic on both ends.