Water, HyClone™ HyPure, Cell Culture Grade

Supplier: Cytiva

SH30529.02 SH30529.LS SH30529.01 SH30529.FS SH30529.03
82007-328EA 19.85 USD
82007-328 16750-114 82007-326 16750-098 82007-330
Water, HyClone™ HyPure, Cell Culture Grade
Water

Eliminate endotoxin concerns with endotoxin-free HyClone Cell Culture-Grade Water, available in a variety of volumes for both research and manufacturing processes.


  • Undergoes extensive purification to meet or exceed the stringent specifications of United States Pharmacopeia (USP)
  • Meets < 0.005 EU/mL endotoxin by turbidimetric assay
  • Manufactured in our ISO 9001 certified facilities


HyClone HyPure Cell Culture Grade Water can be used in multiple cell culture applications. With endotoxin levels as low as < 0.005 EU/mL, HyPure Cell Culture Grade Water supports healthy cell growth and consistent experimental results.


Certifications: Manufactured using ISO 9001 and ISO 13485 certified processes according to 21 CFR 820 medical device current good manufacturing practices (cGMP).

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Endotoxin Concentration < 0.005 EU/mL
Sterilization 0.1 µm sterile filtered
pH 5.0 to 7.0
Storage 2° to 30° C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR