Heidolph® Rotamax 120 Orbital Shaker
Supplier: Heidolph NA, LLC
036130160
82004-958EA 2987.6 USD
82004-958
Heidolph® Rotamax 120 Orbital Shaker
Shakers and Mixers Orbital Shakers
This orbital shaker provides the swirling action needed for the cultivation of microorganisms and cell or tissue cultures.
It is ideal for use with culture plates and Erlenmeyer flasks (≤ 250 ml). Designed for continuous use and features a sparkless and maintenance-free motor. Unit offers timed and continuous operation modes. Process timer allows for unattended operation for up to 120 minutes and sends an acoustic signal upon completion. Enclosed housing ensures virtually no heat transfer from motor to platform, and the unit will self-reset to protect against overheating. Unit also has analog dial control of vibration frequency. The maintenance-free, balanced, sealed drive system is isolated from spills, which makes the shaker ideal for use in cold rooms.
Certifications: Meets the standards of protection class IP 30 (DIN 40050). Tension roller attachment (82004-950) features two rollers to facilitate the shaking of Erlenmeyer flasks.
Ordering information: Manufacturer's three-year unconditional warranty. Unit include a perforated platform and a nonskid rubber mat. For 230V units, contact your Avantor sales representative for more information.