Heidolph® Multi Reax Vibrating Shaker
Supplier: Heidolph NA, LLC
036130040
82004-934EA 3608.8 USD
82004-934
Heidolph® Multi Reax Vibrating Shaker
Shakers and Mixers Orbital Shakers
This versatile shaker can accommodate up to 26 vessels
Unit is designed for continuous use and features a sparkless, maintenance-free motor. It has digital speed control and a self-reset function to protect against overheating. A digital process timer allows for unattended operation for up to 999 minutes and sends an acoustic signal when completed.
Certifications: Meets the standards of protection class IP 30 (DIN 40050).
Ordering information: Manufacturer's three-year unconditional warranty. Unit comes standard with one of each of the carousel attachments. For 230V units, contact your Avantor sales representative for more information.