Heidolph® Reax 20 Overhead Shakers
Supplier: Heidolph NA, LLC
036130400
82004-916EA 8630.11 USD
82004-916 82004-918 82004-914
Heidolph® Reax 20 Overhead Shakers
Shakers and Mixers Orbital Shakers
Units provide gentle overhead movement for wastewater and sludge analysis
Attachment kit 82004-926 consists of four spacers to accommodate four 0.5L bottles. Attachment kit 82004-924 consists of an allen hex wrench and four spacers to accommodate four 1L bottles. The tensioning plate with caps allows for the use of common bottles with a 94mm (311/16") diameter.
They are designed for the easy and fast attachment of bottles and vessels. One-dimensional swirling action is continuously adjustable from 1–16rpm. Units have analog dial control of rpm adjustment. The maintenance-free, balanced, sealed drive system is isolated from spills, which makes these shakers ideal for use in cold rooms. Units are designed for continuous use and will self-reset to protect against overheating.
Certifications: Meet the standards of protection class IP 21 (DIN 40050) and comply with DIN 36414 Item 4 “determination of elutriation with water.”
Ordering information: Manufacturer's three-year unconditional warranty. Units include an attachment for 2L bottles. Other 27cm (105/8") high vessels with a maximum diameter of 13.6cm (53/8") are also compatible. For 230V units, contact your Avantor sales representative for more information.