TELFA® Island Dressings, Sterile, Covidien
Supplier: Cardinal Health
7540-
82004-814CS 727.58 USD
82004-814
TELFA® Island Dressings, Sterile, Covidien
First Aid Supplies Wound Dressings
Convenient, all-in-one dressing has a soft, nonwoven backing, that conforms around wound and seals on all four sides.
It is the ideal dressing for lightly draining wounds. It can also act as a secure dressing for central and peripheral I.V. sites. Nonadherent TELFA® material will not disrupt healing tissue.
Ordering information: Dressings are individually packaged in peel-back packages.