KERLIX™ Super Sponges, Sterile, Covidien
Supplier: Cardinal Health
3085-
82004-770CS 513.62 USD
82004-770
KERLIX™ Super Sponges, Sterile, Covidien
First Aid Supplies Wound Dressings
Prewashed, fluff-dried woven gauze has diamond-fold construction and a crinkle-weave pattern to offer superior loft, bulk, and high absorbency.
This superior primary dressing is designed to transfer exudate away from the wound into the secondary dressing. Absorbency, strength, and low-linting characteristics are ideal for pre-operative prepping, cleansing, debriding, and packing. Sponges are made from high-grade 100% cotton.
Certifications: Exceed USP Type VII gauze requirements.
Ordering information: Supplied in soft pouches, five individually-wrapped sponges per pouch.