Single Bottle Eye Wash Station, Brady®
Supplier: Brady Worldwide
45798
82004-270EA 258.65 USD
82004-270
Single Bottle Eye Wash Station, Brady®
First Aid Supplies Eyewash Stations and Eyewash Bottles
Our single bottle eye wash stations feature a drop down front panel that allows quick access to your eyewash bottles (not included).
  • Stations can be wall or table mounted
Proper usage instructions are conveniently silk-screened on the door. Single bottle station holds one 32 oz. bottle.