Type T Thermocouple Complete System, Kem Scientific
Supplier: J-KEM
210-T-S
82003-728EA 2555.82 USD
82003-728
Type T Thermocouple Complete System, Kem Scientific
Controllers Temperature Controllers
Type T Thermocouple complete system is a new, high speed microprocessor.
  • Efficient power regulation system that adjusts power to the heater 2048 times per second resulting in 0.1 °C regulation
  • USB port with free KEM-Net software enable remote PC control, GLP/GMP compliant data collection
  • KEM-IO feature allows the controller to turn other pieces of equipment On/Off, change the temperature set point
  • Advanced PID algorithm and NIST traceable
Complete system includes one Model 210 temperature controller, one Teflon-coated thermocouple (12" long, 1/8" diameter), one connecting cord and one 14/20 joint adapter.