Simple Green® Industrial Cleaner and Degreaser
Supplier: Janitorial Supplies
SMP13005CT
82003-549CS 297.09 USD
82003-549
Simple Green® Industrial Cleaner and Degreaser
Cleaning Agents Detergents and Surface Cleansers
A powerful all-purpose cleaner and degreaser that provides a safer alternative to toxic cleaners, bleaches and solvents.
  • Orally non-toxic formula
  • Trusted non-flammable, non-abrasive and non-corrosive cleaner is a safer alternative to caustic cleaners and solvents
  • Versatile formula can be used to clean and degrease all washable surfaces
  • Cost-effective concentrate can be used manually or with cleaning equipment
  • VOC compliant and exempt from OSHA and EPA special handling and personal protection requirements for hazardous materials
  • Sassafras scented
The trusted formula is nontoxic, non-abrasive and non-corrosive. Safe for all washable surfaces, Simple Green can be used to remove heavy dirt, grease, food residue and stains from any washable surface including countertops, sinks, tile, carpet, flooring, upholstery and more. The cost-effective concentrate can be used manually or in cleaning equipment such as such as floor scrubbers, dip tanks, carpet extractors, pressure washers, parts washers, steam extractors and foamers. It easily cut through tough grease, oils, cutting fluids, built-up grime and residue on equipment, engines, machinery, vehicles and manufactured parts making it ideal for a wide range of industries including janitorial, manufacturing, MRO, food processing, mining, automotive, transportation and engineering. Butyl and phosphate-free formula.
Ideal for a wide range of industries including manufacturing, MRO, food processing, mining, automotive, transportation and engineering. For carpet, ceramics, cement/concrete, enamel, fabric, fiberglass, laminates, metal, plastic, upholstery.