MetroMax i™ Stem Caster Carts, Metro™
Supplier: Metro
X336EFX3 X366BGX3 X366EGX3 X356EFX3 X356EGX3 X366EFX3 X556EGX3 X566EGX3 X336EGX3 X356BGX3 X536EGX3 X556EFX3 X536EFX3
82003-290EA 1829.49 USD
82003-290 82003-304 82003-308 82003-298 82003-300 82003-306 82003-320 82003-324 82003-332 82003-292 82003-296 82003-312 82003-322 82003-314 82003-288
MetroMax i™ Stem Caster Carts, Metro™
Carts
Carts are ideal for storing and transporting chemicals
Carts feature four shelves that may be adjusted in 1.3 cm (1/2") increments for maximum storage.
Cart shelves feature perimeter ledges to contain spills and safely transport receptacles. Surface ribs keep products from sticking to the shelf. Easy-to-clean shelves and posts are made of strong, durable, reinforced polymers, and are smooth and rounded with no sharp edges. Carts are ideal for use in damp, humid conditions, as well as dry storage. Not recommended for use in temperatures below –29°C (–20°F).
Ordering information: Manufacturer's lifetime warranty against corrosion and rust. Carts are shipped knocked down and can be assembled in minutes without the use of tools. Each cart includes four open-grid or solid-shelf mats, wedge connectors, and four casters (two swivel, two brake) of the indicated type.